4-Chloro-1-benzothiophene-2-carbohydrazide, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

OR12457-1G OR12457-250MG OR12457-5G
77404-314EA 173.82 USD
77404-314 77404-316 77404-318
4-Chloro-1-benzothiophene-2-carbohydrazide, Technical Grade
4-Chloro-1-benzothiophene-2-carbohydrazide
Formula: C₉H₇ClN₂OS
MW: 226.69 g/mol
Melting Pt: 184…186 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09817458
CAS Number: 952182-46-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR