4-Bromo-N-(prop-2-ynyl)benzamide 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR11936-100MG OR11936-1G OR11936-5G
77404-066EA 55.02 USD
77404-066 77404-068 77404-070
4-Bromo-N-(prop-2-ynyl)benzamide 98%
4-Bromo-N-(prop-2-ynyl)benzamide
Formula: C₁₀H₈BrNO
Storage Temperature: Ambient
MDL Number: MFCD09800901
CAS Number: 82225-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR