Methyl-1-butyl-5-oxopyrrolidine-3-carboxylate

Supplier: APOLLO SCIENTIFIC

OR110898-1G OR110898-5G
77403-562EA 220.08 USD
77403-562 77403-564
Methyl-1-butyl-5-oxopyrrolidine-3-carboxylate
Methyl-1-butyl-5-oxopyrrolidine-3-carboxylate
Formula: C₁₀H₁₇NO₃
MW: 199.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD15143238
CAS Number: 59857-87-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR