ethyl-2-((tert-butoxycarbonyl)amino)acetate

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: N-(tert-Butoxycarbonyl)glycine ethyl ester, ethyl 2-((tert-Butoxycarbonyl)amino)acetate, Boc-Gly-OEt, N-Boc-glycine ethyl ester

OR110664-100G OR110664-25G OR110664-5G
77403-398EA 368.82 USD
77403-398 77403-400 77403-402
ethyl-2-((tert-butoxycarbonyl)amino)acetate
ethyl-2-((tert-butoxycarbonyl)amino)acetate
Formula: C₉H₁₇NO₄
Storage Temperature: Ambient
MDL Number: MFCD05663720
CAS Number: 14719-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR