6-Amino-1-heptyl-1H-indazole

Supplier: APOLLO SCIENTIFIC

Synonyms: 1-Heptyl-1H-indazol-6-amine

OR110115-1G OR110115-250MG
77403-086EA 352.52 USD
77403-086 77403-088
6-Amino-1-heptyl-1H-indazole
6-Amino-1-heptyl-1H-indazole
Formula: C₁₄H₂₁N₃
MW: 231.337 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12103247
CAS Number: 1155102-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR