7-Chloro-5-nitro-1H-indazole

Supplier: APOLLO SCIENTIFIC
Warning

OR110736-1G OR110736-250MG
77403-468EA 247.58 USD
77403-468 77403-470
7-Chloro-5-nitro-1H-indazole
7-Chloro-5-nitro-1H-indazole
Formula: C₇H₄ClN₃O₂
Melting Pt: 240…243 °C
Storage Temperature: Ambient
CAS Number: 74209-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR