6-Amino-2H-1,4-benzoxazin-3(4H)-one

Supplier: APOLLO SCIENTIFIC
Warning

OR110727-1G OR110727-5G
77403-462EA 63.16 USD
77403-462 77403-464
6-Amino-2H-1,4-benzoxazin-3(4H)-one
6-Amino-2H-1,4-benzoxazin-3(4H)-one
Formula: C₈H₈N₂O₂
Melting Pt: 266…268 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03425742
CAS Number: 89976-75-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR