6-(tert-Butyl)-4-chlorothieno[3,2-d]pyrimidine 97%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR110628-1G
OR110628-5G
77403-392EA
62.16
USD
77403-392
77403-394
6-(tert-Butyl)-4-chlorothieno[3,2-d]pyrimidine 97%
6-(tert-Butyl)-4-chlorothieno[3,2-d]pyrimidine
Formula:
C₁₀H₁₁ClN₂S MW: 226.726 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03407483 CAS Number: 439693-52-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...