4,5,6,7-Tetrahydro-1-benzothiophene-2-carbaldehyde

Supplier: APOLLO SCIENTIFIC
Warning

OR110615-1G OR110615-250MG
77403-386EA 195.62 USD
77403-386 77403-388
4,5,6,7-Tetrahydro-1-benzothiophene-2-carbaldehyde
4,5,6,7-Tetrahydro-1-benzothiophene-2-carbaldehyde
Formula: C₉H₁₀OS
MW: 166.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16631271
CAS Number: 57021-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR