4-bromo-1-methyl-1H-pyrazole-3-carbonitrile

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Bromo-1-methyl-1H-pyrazole-3-carbonitrile

OR110567-1G OR110567-250MG
77403-380EA 110.04 USD
77403-380 77403-382
4-bromo-1-methyl-1H-pyrazole-3-carbonitrile
4-bromo-1-methyl-1H-pyrazole-3-carbonitrile
Formula: C₅H₄BrN₃
MW: 186.012 g/mol
Melting Pt: 103…107 °C
Storage Temperature: Ambient
MDL Number: MFCD00103545
CAS Number: 287922-71-8
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR