Methyl 2-butyl-1,3-benzoxazole-5-carboxylate

Supplier: APOLLO SCIENTIFIC
Warning

OR110271-1G OR110271-5G
77403-204EA 121.24 USD
77403-204 77403-206
Methyl 2-butyl-1,3-benzoxazole-5-carboxylate
Methyl 2-butyl-1,3-benzoxazole-5-carboxylate
Formula: C₁₃H₁₅NO₃
MW: 233.263 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13182317
CAS Number: 1305712-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR