Ethyl 1-methyl-1H-pyrrole-2-carboxylate

Supplier: APOLLO SCIENTIFIC

OR110264-1G OR110264-5G
77403-196EA 88.64 USD
77403-196 77403-198
Ethyl 1-methyl-1H-pyrrole-2-carboxylate
Ethyl 1-methyl-1H-pyrrole-2-carboxylate
Formula: C₈H₁₁NO₂
Storage Temperature: Ambient
MDL Number: MFCD02179573
CAS Number: 23466-27-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR