2-Methyl-3-nitrobenzenecarbohydrazide

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-Methyl-3-nitrobenzhydrazide

OR110221-10G OR110221-1G OR110221-5G
77403-146EA 424.84 USD
77403-146 77403-148 77403-150
2-Methyl-3-nitrobenzenecarbohydrazide
2-Methyl-3-nitrobenzenecarbohydrazide
Formula: C₈H₉N₃O₃
Storage Temperature: Ambient
MDL Number: MFCD03422927
CAS Number: 869942-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR