4,5-Dibromo-2-(tert-butyl)-3(2H)-pyridazinone

Supplier: APOLLO SCIENTIFIC

OR110076-1G OR110076-5G
77403-066EA 263.88 USD
77403-066 77403-068
4,5-Dibromo-2-(tert-butyl)-3(2H)-pyridazinone
4,5-Dibromo-2-(tert-butyl)-3(2H)-pyridazinone
Formula: C₈H₁₀Br₂N₂O
MW: 310 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12079183
CAS Number: 107784-73-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR