4-(Aminomethyl)tetrahydropyran 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 4-(Aminomethyl)tetrahydro-2H-pyran, 4-Aminomethyltetrahydropyran, Tetrahydro-2H-pyran-4-ylmethanamine, (Tetrahydro-2H-pyran-4-yl)methanamine

OR10333-100G OR10333-25G OR10333-5G
77402-600EA 99.84 USD
77402-600 77402-602 77402-604
4-(Aminomethyl)tetrahydropyran 97%
4-(Aminomethyl)tetrahydropyran
Formula: C₆H₁₃NO
MW: 115.1735 g/mol
Boiling Pt: 75…76 °C/18 mmHg
Density: 1.02 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02179435
CAS Number: 130290-79-8
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR