(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)methanol

Supplier: APOLLO SCIENTIFIC
Warning

OR110037-10G OR110037-1G OR110037-5G
77403-024EA 330.1 USD
77403-024 77403-026 77403-028
(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)methanol
(4-Bromo-3,5-dimethyl-1H-pyrazol-1-yl)methanol
Formula: C₆H₉BrN₂O
Storage Temperature: Ambient
MDL Number: MFCD00085534
CAS Number: 94230-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR