3,4-Dichlorobenzaldehyde 97%

Supplier: APOLLO SCIENTIFIC
Danger

OR10905-100G OR10905-25G
77402-964EA 84.56 USD
77402-964 77402-966
3,4-Dichlorobenzaldehyde 97%
3,4-Dichlorobenzaldehyde
Formula: C₇H₄Cl₂O
MW: 175.01 g/mol
Boiling Pt: 247…248 °C
Melting Pt: 39…42 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003351
CAS Number: 6287-38-3
EINECS: 228-520-5
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR