3-(Aminomethyl)-1H-indole oxalate

Supplier: APOLLO SCIENTIFIC
Warning

OR10823-500MG OR10823-50MG
77402-940EA 137.54 USD
77402-940 77402-942
3-(Aminomethyl)-1H-indole oxalate
3-(Aminomethyl)-1H-indole oxalate
Formula: C₉H₁₀N₂·C₂H₂O₄
MW: 236.224 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01863232
CAS Number: 296775-93-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR