Diethyl [N-(benzylidene)aminomethyl]phosphonate

Supplier: APOLLO SCIENTIFIC
Warning

OR10690-1G OR10690-500MG
77402-882EA 123.28 USD
77402-882 77402-884
Diethyl [N-(benzylidene)aminomethyl]phosphonate
Diethyl [N-(benzylidene)aminomethyl]phosphonate
Formula: C₁₂H₁₈NO₃P
Storage Temperature: Ambient
MDL Number: MFCD00152991
CAS Number: 50917-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR