4-Bromothioanisole

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 1-Bromo-4-(methylthio)benzene, 4-Bromophenyl methyl sulphide

OR10389-1G OR10389-25G OR10389-5G
77402-674EA 21.26 USD
77402-674 77402-676 77402-678
4-Bromothioanisole
4-Bromothioanisole
Formula: C₇H₇BrS
MW: 203.1 g/mol
Boiling Pt: 128…130 °C/9.75 mmHg
Melting Pt: 38…40 °C
Density: 1.49 g/cm³ (25 °C)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00000102
CAS Number: 104-95-0
EINECS: 203-255-8
UN: 3335
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR