Guaiacol 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
1-Hydroxy-2-methoxybenzene, 2-Hydroxyanisole, 2-Methoxyphenol
OR10036-1KG
OR10036-500G
OR10036-5KG
77402-408EA
182.36
USD
77402-408
77402-410
77402-412
Guaiacol 98%
Guaiacol
Formula:
C₇H₈O₂ Boiling Pt: 204…206 °C Melting Pt: 26…31 °C Density: 1.129 g/cm³ (25 °C) Flash Pt: 82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002185 CAS Number: 90-05-1 EINECS: 201-964-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...