(Piperazin-1-yl)(pyrrolidin-1-yl)methanone 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
1-[(Piperazin-1-yl)carbonyl]pyrrolidine, 1-[(Pyrrolidin-1-yl)carbonyl]piperazine
OR0950-1G
OR0950-250MG
OR0950-5G
77402-330EA
78.44
USD
77402-330
77402-332
77402-334
(Piperazin-1-yl)(pyrrolidin-1-yl)methanone 98%
(Piperazin-1-yl)(pyrrolidin-1-yl)methanone
Formula:
C₉H₁₇N₃O MW: 183.251 g/mol Boiling Pt: 106 °C (0.015 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD03844724 CAS Number: 73331-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...