1-(3,4-Dimethoxyphenyl)ethanol

Supplier: APOLLO SCIENTIFIC

OR110953-10G OR110953-1G OR110953-5G
77403-588EA 523.68 USD
77403-588 77403-590 77403-592
1-(3,4-Dimethoxyphenyl)ethanol
1-(3,4-Dimethoxyphenyl)ethanol
Formula: C₁₀H₁₄O₃
Storage Temperature: Ambient
MDL Number: MFCD02127273
CAS Number: 5653-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR