Methyl (2-nitrophenyl)acetate 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR110947-25G OR110947-5G
77403-580EA 80.48 USD
77403-580 77403-582
Methyl (2-nitrophenyl)acetate 98%
Methyl (2-nitrophenyl)acetate
Formula: C₉H₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD00968465
CAS Number: 30095-98-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR