5-Amino-2-diisopropylamino-1,3,4-thiadiazole

Supplier: APOLLO SCIENTIFIC

Synonyms: N,N-Diisopropyl-1,3,4-thiadiazole-2,5-diamine

OR110699-1G OR110699-5G
77403-422EA 176.26 USD
77403-422 77403-424
5-Amino-2-diisopropylamino-1,3,4-thiadiazole
5-Amino-2-diisopropylamino-1,3,4-thiadiazole
Formula: C₈H₁₆N₄S
MW: 200.31 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00036052
CAS Number: 72269-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR