4,5-Dibromothiophene-2-sulfonamide

Supplier: APOLLO SCIENTIFIC

OR110673-1G OR110673-5G
77403-414EA 220.08 USD
77403-414 77403-416
4,5-Dibromothiophene-2-sulfonamide
4,5-Dibromothiophene-2-sulfonamide
Formula: C₄H₃Br₂NO₂S₂
MW: 321.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00208579
CAS Number: 77893-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR