3-Acetamido-4-hydroxybenzoic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: N-(5-Carboxy-2-hydroxyphenyl)acetamide, 2-(Acetylamino)-4-carboxyphenol, 3-(Acetylamino)-4-hydroxybenzoic acid

OR110291-1G OR110291-250MG
77403-238EA 268.96 USD
77403-238 77403-240
3-Acetamido-4-hydroxybenzoic acid
3-Acetamido-4-hydroxybenzoic acid
Formula: C₉H₉NO₄
Melting Pt: 238…241 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD14583147
CAS Number: 91004-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR