1,3-Benzothiazole-2-carbohydrazide

Supplier: APOLLO SCIENTIFIC
Warning

OR110285-5G OR110285-10G OR110285-1G
77403-234EA 209.88 USD
77403-234 77403-230 77403-232
1,3-Benzothiazole-2-carbohydrazide
1,3-Benzothiazole-2-carbohydrazide
Formula: C₈H₇N₃OS
Melting Pt: 179 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02713577
CAS Number: 28891-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR