3-Pyridineboronic acid pinacol ester

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Pyridine-3-boronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

OR11024-100G OR11024-25G OR11024-5G
77403-168EA 140.6 USD
77403-168 77403-170 77403-176
3-Pyridineboronic acid pinacol ester
3-Pyridineboronic acid pinacol ester
Formula: C₁₁H₁₆BNO₂
Melting Pt: 102…104 °C
Storage Temperature: Ambient
MDL Number: MFCD03084758
CAS Number: 329214-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR