Chlorotrimethylsilane 99%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
TMCS, TMSC, Trimethylchlorosilane
OR10000-1L
OR10000-250ML
77402-386EA
74.38
USD
77402-386
77402-388
Chlorotrimethylsilane 99%
Chlorotrimethylsilane
Formula:
C₃H₉ClSi MW: 108.64 g/mol Boiling Pt: 56…58 °C Melting Pt: ca …58 °C Density: 0.856 g/cm³ (25 °C) Flash Pt: –18 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000502 CAS Number: 75-77-4 EINECS: 200-900-5 UN: 1298 ADR: 3+8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...