3-Methylpicolinic acid

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-Methylpyridine-2-carboxylic acid

OR0922-1G OR0922-5G
77402-312EA 21.26 USD
77402-312 77402-314
3-Methylpicolinic acid
3-Methylpicolinic acid
Formula: C₇H₇NO₂
MW: 137.14 g/mol
Melting Pt: 114…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00191553
CAS Number: 4021-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR