(Morpholin-4-yl)(piperazin-1-yl)methanone 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR0241-1G OR0241-250MG
77401-962EA 68.64 USD
77401-962 77401-964
(Morpholin-4-yl)(piperazin-1-yl)methanone 98%
(Morpholin-4-yl)(piperazin-1-yl)methanone
Formula: C₉H₁₇N₃O₂
Melting Pt: 85…88 °C
Storage Temperature: Ambient
MDL Number: MFCD03844725
CAS Number: 98834-08-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR