6,8-Dibromo-5-chloroquinazolin-4-ol

Supplier: APOLLO SCIENTIFIC

OR111102-1G OR111102-250MG
77403-650EA 220.08 USD
77403-650 77403-652
6,8-Dibromo-5-chloroquinazolin-4-ol
6,8-Dibromo-5-chloroquinazolin-4-ol
Formula: C₈H₃Br₂ClN₂O
MW: 338.383 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28954638
CAS Number: 1858241-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR