6-Chloro-2-methyl-4-pyrimidinol

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 6-Chloro-4-hydroxy-2-methylpyrimidine, 6-Chloro-2-methylpyrimidin-4(1H)-one

OR110379-1G OR110379-5G
77403-296EA 131.42 USD
77403-296 77403-298
6-Chloro-2-methyl-4-pyrimidinol
6-Chloro-2-methyl-4-pyrimidinol
Formula: C₅H₅ClN₂O
MW: 144.558 g/mol
Melting Pt: 206…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00030773
CAS Number: 17551-52-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR