tert-Butyl (diethylphosphono)acetate 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
Synonyms:
tert-Butyl 2-(diethoxyphosphoryl)acetate, Diethyl(2-tert-butoxycarbonylmethyl)phosphonate
OR10694-500G
OR10694-100G
OR10694-1KG
77402-894EA
241.46
USD
77402-894
77402-890
77402-892
tert-Butyl (diethylphosphono)acetate 98%
tert-Butyl (diethylphosphono)acetate
Formula:
C₁₀H₂₁O₅P MW: 252.25 g/mol Boiling Pt: 100…103 °C/1.5 mmHg Density: 1.074 g/cm³ (25 °C) Flash Pt: >113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00075414 CAS Number: 27784-76-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...