3-Phenylbenzoic acid 98+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: Biphenyl-3-carboxylic acid, 3-Biphenylcarboxylic acid, 1,1'-Biphenyl-3-carboxylic acid, [1,1'-Biphenyl]-3-carboxylic acid

OR10604-1G OR10604-25G OR10604-5G
77402-818EA 21.26 USD
77402-818 77402-820 77402-822
3-Phenylbenzoic acid 98+%
3-Phenylbenzoic acid
Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
Melting Pt: 164…168 °C
Storage Temperature: Ambient
MDL Number: MFCD00045846
CAS Number: 716-76-7
UN: 3077
ADR: 9+EHS,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR