2,3,3-Trimethylindolenine

Supplier: APOLLO SCIENTIFIC
Warning

OR10172-100G OR10172-25G OR10172-500G
77402-514EA 57.06 USD
77402-514 77402-516 77402-518
2,3,3-Trimethylindolenine
2,3,3-Trimethylindolenine
Formula: C₁₁H₁₃N
MW: 159.23 g/mol
Boiling Pt: 228…229 °C/744 mmHg
Density: 0.992 g/cm³ (25 °C)
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00005724
CAS Number: 1640-39-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR