Homopiperazine 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 1,4-Diazepane, 1,4-Diazacycloheptane, Perhydro-1,4-diazepine

OR0503-100G OR0503-25G
77402-104EA 96.78 USD
77402-104 77402-106
Homopiperazine 97%
Homopiperazine
Formula: C₅H₁₂N₂
MW: 100.16 g/mol
Boiling Pt: 168…170 °C
Melting Pt: 38…40 °C
Density: 0.95 g/cm³ (25 °C)
Flash Pt: 61 °C
Storage Temperature: Ambient
MDL Number: MFCD00006933
CAS Number: 505-66-8
EINECS: 208-016-1
UN: 3259
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR