2,6-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR0482-1G
OR0482-250MG
77402-096EA
393.28
USD
77402-096
77402-098
2,6-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid
2,6-Dimethylimidazo[1,2-a]pyridine-3-carboxylic acid
Formula:
C₁₀H₁₀N₂O₂ MW: 190.198 g/mol Melting Pt: 165…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD00269372 CAS Number: 81438-52-0 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...