(4-Pyridylmethyl)hydrazine dihydrochloride 95+%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-(Hydrazinylmethyl)pyridine dihydrochloride, 4-(Hydrazinomethyl)pyridine dihydrochloride

OR0207-1G OR0207-250MG
77401-946EA 65.88 USD
77401-946 77401-948
(4-Pyridylmethyl)hydrazine dihydrochloride 95+%
(4-Pyridylmethyl)hydrazine dihydrochloride
Formula: C₆H₁₁Cl₂N₃
Melting Pt: >200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD06245508
CAS Number: 89598-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR