2-(3,5-Dichlorophenyl)hydrazinecarbothioamide, Technical Grade

Supplier: APOLLO SCIENTIFIC
Warning

OR111045-10G OR111045-1G OR111045-5G
77403-620EA 883.6 USD
77403-620 77403-622 77403-624
2-(3,5-Dichlorophenyl)hydrazinecarbothioamide, Technical Grade
2-(3,5-Dichlorophenyl)hydrazinecarbothioamide
Formula: C₇H₇Cl₂N₃S
MW: 236.122 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03305525
CAS Number: 96423-39-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR