5-Methoxy-1H-indazole

Supplier: APOLLO SCIENTIFIC

OR110371-1G OR110371-250MG
77403-286EA 113.1 USD
77403-286 77403-288
5-Methoxy-1H-indazole
5-Methoxy-1H-indazole
Formula: C₈H₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD07781657
CAS Number: 94444-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR