Tetrabutylammonium bromide 98%
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR10054-100G
OR10054-500G
77402-422EA
24.46
USD
77402-422
77402-424
Tetrabutylammonium bromide 98%
Tetrabutylammonium bromide
Formula:
C₁₆H₃₆BrN Melting Pt: 101…106 °C Density: 1.15 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011633 CAS Number: 1643-19-2 EINECS: 216-699-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...