Ethyl 3-(chlorosulfonyl)isonicotinate 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Ethyl 3-(chlorosulphonyl)pyridine-4-carboxylate

OR0448-250MG OR0448-50MG
77402-090EA 294.58 USD
77402-090 77402-092 77402-094
Ethyl 3-(chlorosulfonyl)isonicotinate 95%
Ethyl 3-(chlorosulfonyl)isonicotinate
Formula: C₈H₈ClNO₄S
MW: 249.671 g/mol
Boiling Pt: 72…78 °C (0.1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD01313855
CAS Number: 306936-12-9
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR