1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

OR0419-1G OR0419-250MG
77402-076EA 63.6 USD
77402-076 77402-078 77402-080
1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride
1,3-Dimethyl-1H-pyrazole-5-carbonyl chloride
Formula: C₆H₇ClN₂O
Boiling Pt: 100 °C/20 mmHg
Storage Temperature: Refrigerator
MDL Number: MFCD00833258
CAS Number: 55458-67-8
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR