3-(4'-Chlorophenyl)propanol 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 3-(4-Chlorophenyl)propan-1-ol

OR0287-1G OR0287-25G OR0287-5G
77401-998EA 21.4 USD
77401-998 77402-000 77402-002
3-(4'-Chlorophenyl)propanol 95%
3-(4'-Chlorophenyl)propanol
Formula: C₉H₁₁ClO
Boiling Pt: 98…102 °C/0.1 mmHg
Storage Temperature: Ambient
MDL Number: MFCD00831102
CAS Number: 6282-88-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR