6-Amino-3-chloro-1H-indazole

Supplier: APOLLO SCIENTIFIC
Warning

OR110175-1G OR110175-5G
77403-106EA 55.02 USD
77403-106 77403-108
6-Amino-3-chloro-1H-indazole
6-Amino-3-chloro-1H-indazole
Formula: C₇H₆ClN₃
Storage Temperature: Refrigerator
MDL Number: MFCD07781648
CAS Number: 21413-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR