2-Chloro-4-methylphenylboronic acid
Supplier: APOLLO SCIENTIFIC
|
Warning
|
OR10414-1G
OR10414-25G
OR10414-5G
77402-706EA
21.26
USD
77402-706
77402-708
77402-710
2-Chloro-4-methylphenylboronic acid
2-Chloro-4-methylphenylboronic acid
Formula:
C₇H₈BClO₂ Melting Pt: 198…202 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD03411936 CAS Number: 145349-62-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...