5-Bromo-2-chlorobenzoic acid 97+%

Supplier: APOLLO SCIENTIFIC
Warning

OR1038-100G OR1038-25G OR1038-5G
77402-660EA 57.06 USD
77402-660 77402-668 77402-670
5-Bromo-2-chlorobenzoic acid 97+%
5-Bromo-2-chlorobenzoic acid
Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
Melting Pt: 157…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00002415
CAS Number: 21739-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR