1,3-Dimethyl-1H-pyrazole-5-carboxylic acid 98%

Supplier: APOLLO SCIENTIFIC
Warning

OR0417-25G OR0417-5G
77402-072EA 172.18 USD
77402-072 77402-074
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid 98%
1,3-Dimethyl-1H-pyrazole-5-carboxylic acid
Formula: C₆H₈N₂O₂
MW: 140.1400 g/mol
Melting Pt: 206…213 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00159636
CAS Number: 5744-56-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR